EMPO 95 HPLC_ASSAY_METHOD

Supplier: Enzo Life Sciences

Synonyms: 2-Ethoxycarbonyl-2-methyl-3,4-dihydro-2H-pyrrole-1-oxide

430-098-M010 430-098-M050
89151-106EA 162.74 CAD
89151-106 89151-108
EMPO 95 HPLC_ASSAY_METHOD
EMPO

Most efficient spin trap for the in vitro and in vivo detection of O-, N-, S-, and C-centered free radicals.


  • Formulation: Colorless to light yellow oil.


These having a longer life-time than DMPO.


Hygroscopic and soluble in water, DMSO, ethanol or dichloromethane.

Formula: C₈H₁₃NO₃
MW: 171.2 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 61856-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0b527b360d80e6f18be1bb72ffa462f33da4c6e&vid=3634ef8c0deb65014ad3c9182d0f723a926fa793&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (NMR,HPLC) ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR