Wogonin ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: 5,7-Dihydroxy-8-methoxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxy-8-methoxyflavone

385-033-M025
89150-550EA 141 CAD
89150-550 89150-552
Wogonin ≥98% (by HPLC)
Wogonin

Anti-inflammatory and antioxidant.


  • Formulation: White to yellow solid.


Cell permeable and orally available flavonoid. Induces apoptosis in tumor cells but not in healthy cells by increasing peroxide production. Displays anti-inflammatory properties. Suppresses the release of nitric oxide (NO) by inducible nitric oxide synthase (iNOS; NOS II), PGE2 by cyclooxygenase-2 (COX-2), proinflammatory cytokines, MCP-1 gene expression and NF-κB activation.


Soluble in DMSO (10 mg/mL).


Synonym: 5,7-Dihydroxy-8-methoxyflavone

Formula: C₁₆H₁₂O₅
MW: 284.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017736
CAS Number: 632-85-9
UN: 2790
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50948999b9842512760ee1136ae289e7198dc760&vid=db6a6d40c5199bf29502429b019f78ce4e25eb2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by ¹H-NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR