Ciprofloxacin hydrochloride monohydrate ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 1-Cyclopropyl- 6-Fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride monohydrate, 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride monohydrate, Nanomycopulitine

380-288-G005
89150-362EA 74.26 CAD
89150-362
Ciprofloxacin hydrochloride monohydrate ≥98% (by HPLC)
Ciprofloxacin hydrochloride monohydrate

Fluoroquinolone antibacterial agent


  • Effective against Mycobacterium tuberculosis
  • Potentially anxiogenic


Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria.


White to light yellow powder


Soluble in water (35 mg/ml)


Packaging: Shipping temperature: Ambient

Formula: C₁₇H₁₈FN₃O₃·HCl·H₂O
MW: 385.82 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00242856
CAS Number: 86393-32-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3475dfbfae56a28192995bdd7cb99cef0c8f22d1&vid=6590997454a5eeb431e63ffe7ae4b80cef703d60&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR