Ampicillin sodium salt ≥93%

Supplier: Enzo Life Sciences

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

380-268-G005
89150-308EA 109.02 CAD
89150-308
Ampicillin sodium salt ≥93%
Ampicillin sodium salt

β-Lactam antibiotic


  • Semisynthetic from penicillin
  • Inactivated by β-lactamase


Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking). Inactivates transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-negative and Gram-positive bacteria.


White to light yellow powder


Soluble in water (50 mg/mL)


Packaging: Shipping temperature: Ambient

Formula: C₁₆H₁₈N₃NaO₄S
Storage Temperature: Freezer
MDL Number: MFCD00064313
CAS Number: 69-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fdaf51b3eba4e947a5c5c6f0204ee44520cd69b&vid=852ee2c1eabb7f1ded7197a6a9203635619a9e2c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR