Rifampicin ≥97%

Supplier: Enzo Life Sciences

380-071-M100
89149-944EA 72.68 CAD
89149-944
Rifampicin ≥97%
Rifampicin

Bacterial DNA-dependent RNA polymerase inhibitor


A selective inhibitor of bacterial DNA-dependent RNA polymerase (RNAP). Has no effect on mammalian RNA polymerase. The RNAP-rifampicin complex locks the promoter in an abortive initiation reaction making short oligoribonucleotides which diffuse out of the active site. Inhibits alpha-synuclein fibrillation and disaggregates fibrils. Potent inducer of hepatic cytochrome P450 enzymes.


Semisynthetic.

Formula: C₄₃H₅₈N₄O₁₂
MDL Number: MFCD00151389
CAS Number: 13292-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e104455ebe3805c5a2a7cd538a05950cfd23b945&vid=4280f7aeba5dead4d1e4df122cfcd0f6133cb732&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR