Daunorubicin hydrochloride (from Streptomyces species)

Supplier: Enzo Life Sciences

Synonyms: Daunorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-8-Acetyl-10-(((2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

380-043-M010
89149-886EA 285.98 CAD
89149-886
Daunorubicin hydrochloride (from Streptomyces species)
Daunorubicin hydrochloride

Antitumor antibiotic.


Anthracycline anticancer antibiotic. Induces DNA damage by intercalating into DNA and inhibiting topoisomerase I and II. Inhibits RNA and DNA synthesis. Induces DNA single strand breaks and apoptosis in tumor cells.


Orange to red crystalline powder


Soluble in water or methanol


Packaging: Shipping temperature: Ambient

Formula: C₂₇H₂₉NO₁₁·HCl
MW: 563.98 g/mol
Storage Temperature: Freezer
MDL Number: MFCD04974507
CAS Number: 23541-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4affb0909af92d9ee03a31eb5a94b418d62f8699&vid=fe9db3822c999c07838290ae3ea9f5f2a77d1828&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR