Fusidate sodium ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: Fusidate Sodium, Fusidic acid sodium, Sodium (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoate

380-011-M050
89149-804EA 72.68 CAD
89149-804
Fusidate sodium ≥98% (by TLC)
Fusidate sodium

Antibiotic which suppresses nitric oxide (NO) toxicity in pancreatic islet cells. Inhibits protein synthesis by inhibition of elongation factor G at the level of the ribosome.


  • Elongation factor G inhibitor


White to off-white solid.


Soluble in water (50mg/ml).

Formula: C₃₁H₄₇NaO₆
MW: 538.701 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09054714
CAS Number: 751-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea7c507dceed6db0744e661622afdd031e118753&vid=9d82ccf720218d848af508c82a3358368649e63e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR