D-erythro-Sphingosine (synthetic) ≥99%

Supplier: Enzo Life Sciences

Synonyms: D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol

306-011-M025
89148-704EA 148.52 CAD
89148-704
D-erythro-Sphingosine (synthetic) ≥99%
D-erythro-Sphingosine

PKC inhibitor


Common component of cell membranes, often as phosphatidyl conjugates. Inhibitor of protein kinase C (PKC) and calmodulin-dependent enzymes.

Formula: C₁₈H₃₇NO₂
MW: 299.50 g/mol
MDL Number: MFCD00036751
CAS Number: 123-78-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c2468f54513fc2ae45d1ac5474d92f84d93ac75&vid=23f0cb5098717172dc27bfb363b3bc8afcb73f98&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR