Imatinib mesylate

Supplier: Enzo Life Sciences

Synonyms: N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulphonate, N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulfonate, Imatinib Methanesulphonate, Imatinib Methanesulfonate

270-492-M500
89148-082EA 225.01 CAD
89148-082 89148-084
Imatinib mesylate
Imatinib mesylate

Tyrosine kinase inhibitor


Tyrosine kinase inhibitor. Highly specific for BCR-ABL, the enzyme associated with chronic myelogenous leukemia (CML) and certain forms of acute lymphoblastic leukemia (ALL). Also shown to inhibit the transmembrane receptor kit and platelet-derived growth factor (PDGF) receptors.

Formula: C₂₉H₃₁N₇O·CH₄SO₃
MW: 589.72 g/mol
MDL Number: MFCD04307699
CAS Number: 220127-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c279d3d3260c5978d1e536b10cee8d573d36e37&vid=c4f8d33c9e3c0cae97c7b7195ea932baa6de05aa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR