Fenofibrate ≥98%

Supplier: Enzo Life Sciences
Warning

Synonyms: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate

270-481-G005
89148-054EA 80.58 CAD
89148-054
Fenofibrate ≥98%
Fenofibrate

PPARα activator.


  • Formulation: White to off-white crystalline solid.


Hypolipidemic compound. PPARα (peroxisome proliferator-activated receptor α) agonist (EC₅₀ = 18 µM for mouse and EC₅₀ = 30 µM for human PPARα). Also binds to PPARγ, but with at least 10-fold less affinity and is inactive at PPARδ (up to EC₅₀ = 100 µM).


Soluble in acetone, ether, DMSO or dimethyl formamide; slightly soluble in 100% ethanol or methanol; insoluble in water.


Synonyms: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methyl-propanoic acid 1-methylethyl ester, procetofen

Formula: C₂₀H₂₁ClO₄
MW: 360.83 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00133314
CAS Number: 49562-28-9
EINECS: 256-376-3
Merck Index: 14,03978
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc2e18ed8980bab97534e02358c36f0c568cbcfc&vid=1924bebf1b044dd1af3c1f7450900f94f1d22c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR