Ciprofibrate ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

270-475-M025
89148-026EA 110.6 CAD
89148-026 89148-028
Ciprofibrate ≥98% (by HPLC)
Ciprofibrate

PPARα activator.


  • Formulation: White to light yellow powder.


Hypolipidemic compound. PPARα (peroxisome proliferator-activated receptor α) agonist.


Soluble in DMSO.


Synonym: 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid

Formula: C₁₃H₁₄Cl₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00467135
CAS Number: 52214-84-3
EINECS: 257-744-6
Merck Index: 14,02313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b69ed610e5881f1171311ae0c9f6a8264f919d0&vid=60019995356404c9bcdd7217506bd4c6cd560287&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR