AS-604850 ≥98% (by HPLC and UV method)

Supplier: Enzo Life Sciences

270-461-M005
89147-972EA 290.72 CAD
89147-972
AS-604850 ≥98% (by HPLC and UV method)
AS-604850

Potent, cell permeable and ATP-competitive inhibitor of phosphoinositide 3-kinase γ (PI(3)Kγ).


Exhibits selectivity over other PI(3)K isoforms.

Formula: C₁₁H₅F₂NO₄S
MDL Number: MFCD19705528
CAS Number: 648449-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8306c24f2b4cc3f5876b9e7a22442f841f8235d&vid=8336e283c459f801f2f43081357a6c6cc200b816&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR