LY 303511 ≥95% (by HPLC)

Supplier: Enzo Life Sciences

270-410-M001 270-410-M005
89147-820EA 142.2 CAD
89147-820 89147-822
LY 303511 ≥95% (by HPLC)
LY 303511

Useful negative control for the PI(3)K (phosphoinositide 3-kinase) inhibitor LY-294,002. Does not inhibit PI(3)K even at concentrations


Yellow solid.


Soluble in DMSO.

Formula: C₁₉H₁₈N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD03453556
CAS Number: 154447-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d49c5c5579839ee94f8459eab400c783f1ad9a1&vid=d33f8c76d9d1e17addc5ed4c2bd0309831f58c87&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR