Tyrphostin AG 1024 ≥98% (by HPLC and UV method)

Supplier: Enzo Life Sciences

270-217-M001 270-217-M005
89147-414EA 170.64 CAD
89147-414 89147-416
Tyrphostin AG 1024 ≥98% (by HPLC and UV method)
Tyrphostin AG 1024

Specific inhibitor of insulin-like growth factor-1 (IGF-1) and insulin receptor kinases. Induces apoptosis by downregulation of the expression of phospho-Akt1 and Bcl-2. Also increases Bax, p53 and p21 expression.


  • IGF-1 inhibitor


Yellow powder.


Soluble in acetonitrile (5mg/ml), DMSO (8mg/ml), dichloromethane (23mg/ml) or methanol (5mg/ml); insoluble in 100% ethanol.

Formula: C₁₄H₁₃BrN₂O
Storage Temperature: Freezer
MDL Number: MFCD02179365
CAS Number: 65678-07-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91b0cd9ee24ec9a6d2382a42c8328b99546b6718&vid=2d00a7fbe650f35440f7aca11e10922c333095d0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR