Piceatannol ≥98%

Supplier: Enzo Life Sciences

270-202-M001 270-202-M010 270-202-M050
89147-358EA 79 CAD
89147-358 89147-360 89147-362 89147-364
Piceatannol ≥98%
Piceatannol

Syk inhibitor


Originally isolated from Euphorbia lagascae. Selective protein tyrosine kinase Syk inhibitor (IC50~10 µM). In RBL-2H3 cells the selective inhibition of Syk by piceatannol results in inhibition of FcεR1-mediated signaling. Activator of human deacetylase SIRT1.


Synthetic.


SIRT1 activator.

Formula: C₁₄H₁₂O₄
MDL Number: MFCD00221715
CAS Number: 10083-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7e7ac33c5a57426b351db576f7db5e4d10190ae&vid=dae605b83d39130efe5e831acde76ff52ed63c85&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR