Anti-TLR5 Goat Polyclonal Antibody

Supplier: Enzo Life Sciences
210-853-C200
89146-166EA 750.5 CAD
89146-166
Anti-TLR5 Goat Polyclonal Antibody
Antibodies
Note: Human Toll-like Receptor 5 (TLR5) is also described as Toll/Interleukin-1 Receptor-like Gene 3 (TIL3).

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human TLR5.

Type: Primary
Antigen: TLR5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR