p70 S6K Activity Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-EKS-470
89141-850EA 1265.58 CAD
89141-850
p70 S6K Activity Kit, Enzo Life Sciences
Assays Protein Assays
For the measurement of p70 S6K activity in partially purified, purified, or crude enzyme preparations from any species.
The p70 S6K activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2