Anti-Nitrotyrosine Mouse Monoclonal Antibody
Supplier: Enzo Life Sciences
905-763-100
89141-426EA 673.08 CAD
89141-426
Anti-Nitrotyrosine Mouse Monoclonal Antibody
Antibodies
The enzymatic conversion of arginine to citrulline by nitric oxide synthase (NO Synthase) leads to the formation of nitric oxide (NO). Peroxynitrite is formed following reaction of NO with superoxide, producing a potent oxidant that reacts with a variety of biological molecules, including tyrosine residues in proteins. Tyrosine nitration can inactivate enzymes and receptors that depend on tyrosine residues for their activity. Nitration prevents phosphorylation of tyrosine residues important for signal transduction.
  • For ELISA, Flow Cytometry, Western Blot
Recognizes human, mouse,More Product Information