Agarose, Pulse Field Gel Electrophoresis, IBI Scientific
Supplier: IBI Scientific
IB70060 IB70061 IB70062 IB70063 IB70064 IB70065
89133-108EA 67.02 CAD
89133-108 89133-110 89133-112 89133-114 89133-116 89133-118
Agarose, Pulse Field Gel Electrophoresis, IBI Scientific
Electrophoresis Gels Agarose and Acrylamide
Pulse Field Gel Electrophoresis Agarose is a linear polymer with a high molecular weight, creating gel structures unlike those of standard agarose
Some features of IBI PFGE Agarose are extrememly high gel strength, greater thermal stability when compared to standard agarose, and exceptionally low absorption of staining reagents.
IBI PFGE Agarose can be used for pulse field gel electrophoresis, conventional electrophoresis and blotting, and cell and enzyme immobilization. PFGE Agarose will separate and finely resolve molecules > 1000bp.
This characteristic, along with its very lowMore Product Information