LabExact® Grade VSS 1.5 µm Binderless Glass Microfiber Filters
Supplier: I.W. TREMONT CO., INC.
LEVSS3700 LEVSS4250 LEVSS4700 LEVSS5500 LEVSS7000 LEVSS8260 LEVSS9000 LEVSSS810 LEVSS1020 LEVSS1100 LEVSS1250 LEVSS1500 LEVSS2100 LEVSS2400 LEVSS2500 LEVSS3200 LEVSS3500
77591-686BX 34.13 CAD
77591-686 77591-688 77591-690 77591-692 77591-694 77591-696 77591-698 77591-700 77591-668 77591-670 77591-672 77591-674 77591-676 77591-678 77591-680 77591-682 77591-684
LabExact® Grade VSS 1.5 µm Binderless Glass Microfiber Filters
Filters Glass Fiber Filters
LabExact® Grade VSS is a binder-free material manufactured using a proprietary glass chemistry which permits usage in high heat applications beyond typical borosilicate glass blends. Ideally suited for determination of fixed and volatile solids ignited at 550 °C Method 2540E.
  • Particle retention 1.50 µm and high loading capacity is an attribute of the high surface area and complex pore structure
  • The residue of TSS, TDS after heating to dryness for a defined amount of time and at a defined temperature is defined as Fixed solids
  • Volatile solids are those
More Product Information