LabExact® Grade TSS 1.5 µm Binderless Glass Microfiber Filters
Supplier: I.W. TREMONT CO., INC.
LETSS1020 LETSS1100 LETSS1250 LETSS1500 LETSS2100 LETSS2400 LETSS2500 LETSS3200 LETSS3500 LETSS3700 LETSS4250 LETSS4700 LETSS5500 LETSS7000 LETSS8260 LETSS9000 LETSSS810
77591-626BX 80.58 CAD
77591-626 77591-628 77591-630 77591-632 77591-634 77591-636 77591-638 77591-640 77591-642 77591-644 77591-646 77591-648 77591-650 77591-652 77591-654 77591-656 77591-658
LabExact® Grade TSS 1.5 µm Binderless Glass Microfiber Filters
Filters Glass Fiber Filters
LabExact® Grade TSS is a binderless high efficiency (HEPA type) filter mediµm with particle retention of 1.50µm. Liquid filtration properties have been specifically designed for EPA Methods 2540C and 2540D for testing dissolved and suspended solids in water and wastewater.
  • High flow rate with high capacity
  • Binderless borosilicate glass fiber media has no added extractables to aid in the elimination of sample contamination
  • Excellent wet strength
Binderless borosilicate glass microfiber high efficiency (HEPA type) filter mediµm. Excellent wet strengthMore Product Information