LabExact® Grade D 2.7 µm Binderless Glass Microfiber Filters
Supplier: I.W. TREMONT CO., INC.
LED1100 LED1250 LED1500 LED2100 LED2400 LED2500 LED3500 LED3700 LED4250 LED4700 LED5500 LED7000 LED9000
77590-890BX 172.36 CAD
77590-890 77590-892 77590-894 77590-896 77590-898 77590-900 77590-902 77590-904 77590-906 77590-908 77590-910 77590-912 77590-914
LabExact® Grade D 2.7 µm Binderless Glass Microfiber Filters
Filters Glass Fiber Filters
LabExact® Grade D Coarse porosity, fast flow rate, with a 2.7 µm size particle retention. Well suited for higher volume and repetitive laboratory filtering. Also ideally used as a general prefilter to extend primary filter life.
  • Glass microfiber filters are produced from 100% borosilicate glass microfiber without the use of binders or strengthening agents
  • Depth filtration categorizes filter media that traps particulate within the random matrix of fibers found within the thickness of the media
  • The efficiency rating of the filter is characteristic of the
More Product Information