LabExact® Grade A 1.6 µm Binderless Glass Microfiber Filters
Supplier: I.W. TREMONT CO., INC.
LEA3500 LEA3700 LEA4250 LEA4700 LEA5500 LEA7000 LEA9000 LEA1100 LEA1250 LEA1500 LEA2100 LEA2400 LEA2500
77590-346BX 31.6 CAD
77590-346 77590-348 77590-350 77590-352 77590-354 77590-356 77590-386 77590-334 77590-336 77590-338 77590-340 77590-342 77590-344
LabExact® Grade A 1.6 µm Binderless Glass Microfiber Filters
Filters Glass Fiber Filters
LabExact® Grade A Fine porosity, fast flow rate, with a 1.6 µm size particle retention. Frequently used in the filtration of precipitated proteins and cells. Ideally suited for use as a filter for radioimmuno assay of weak beta emitters by scintillation counting and gravimetric determination of airborne particulate.
  • Glass microfiber filters are produced from 100% borosilicate glass microfiber, without the use of binders or strengthening agents
  • Depth filtration categorizes filter media that traps particulate within the random matrix of fibers found within
More Product Information