Human Aminoadipate Aminotransferase (AADAT) ELISA Kit

Supplier: Bioss

BSKH63025-48T
77537-720EA 729.69 CAD
77537-720
Human Aminoadipate Aminotransferase (AADAT) ELISA Kit
Assays ELISAs

  • High sensitivity with a broad assay range
  • Flexible inputs: Serum, plasma, cell culture supernatant
  • Reproducible and specific results

New Product

Order Now

SPECIFICATIONS

Environmentally Preferable
Target protein Aminoadipate Aminotransferase (AADAT)
Primary antibody reactivity Human
Assay Type Sandwich
Host
Conjugate ELISA Biotin
Format Pre-coated
Assay duration Multiple steps
Sensitivity 0.062 ng/ml
Detection range 0.156 - 10 ng/ml
Detection method Colorimetric microplate reader primary wavelength 450 nm
Storage temperature TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
Shelf life 12 Months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR