Anti-eNOS Ser1176 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-13071R
77440-410EA 601.4 CAD
77440-410
Anti-eNOS Ser1176 Rabbit Polyclonal Antibody
Antibodies
eNOS is a calcium/calmodulin dependent enzyme which undergoes several post translational modifications, including acylation with myristate and palmitate, and phosphorylation on numerous residues. As with the other members of the NOS family, eNOS derives the diffusible multifunctional second messenger NO from L arginine through a series of reactions in which L citrulline is an intermediate. eNOS plays an important role in controlling vascular tone, platelet aggregation, and cardiac myocyte function.

Type: Primary
Antigen: eNOS Ser1176
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR