Anti-15 Lipoxygenase 1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-24356R
77440-160EA 601.4 CAD
77440-160
Anti-15 Lipoxygenase 1 Rabbit Polyclonal Antibody
Antibodies
Oxygenase and 14,15-leukotriene A4 synthase activity. Converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. Also acts on C-12 of arachidonate as well as on linoleic acid.

Type: Primary
Antigen: 15 Lipoxygenase 1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR