Anti-Asparagine Synthetase Rabbit Polyclonal Antibody

Supplier: Bioss

BS-23431R
77439-964EA 601.4 CAD
77439-964
Anti-Asparagine Synthetase Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is involved in the synthesis of asparagine. This gene complements a mutation in the temperature-sensitive hamster mutant ts11, which blocks progression through the G1 phase of the cell cycle at nonpermissive temperature. Alternatively spliced transcript variants have been described for this gene. [provided by RefSeq, May 2010].

Type: Primary
Antigen: Asparagine synthetase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR