Anti-SLIRP/C14orf156 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-21033R
77439-338EA 601.4 CAD
77439-338
Anti-SLIRP/C14orf156 Rabbit Polyclonal Antibody
Antibodies
Steroid receptor RNA activator (SRA, or SRA1; MIM 603819) is a complex RNA molecule containing multiple stable stem-loop structures that functions in coactivation of nuclear receptors. SLIRP interacts with stem-loop structure-7 of SRA (STR7) and modulates nuclear receptor transactivation (Hatchell et al., 2006 (PubMed 16762838)).(supplied by OMIM, Mar 2008).

Type: Primary
Antigen: SLIRP/C14orf156
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen name SLIRP/C14orf156
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Gene ID 81892
Isotype IgG (H, L)
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Molecular weight 12
Purification Purified by protein A
Concentration 1 µg/µl
Storage buffer 0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
Storage temperature Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR