Anti-NAPE-PLD Rabbit Polyclonal Antibody

Supplier: Bioss

BS-19015R
77438-578EA 601.4 CAD
77438-578
Anti-NAPE-PLD Rabbit Polyclonal Antibody
Antibodies
NAPEPLD is a phospholipase D type enzyme that catalyzes the release of N-acylethanolamine (NAE) from N-acyl-phosphatidylethanolamine (NAPE) in the second step of the biosynthesis of N-acylethanolamine (Okamoto et al., 2004 (PubMed 14634025)).(supplied by OMIM, Oct 2008).

Type: Primary
Antigen: NAPE-PLD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR