Anti-CYP 3A4, 3A7, 3A5, and 3A9 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-1742R
77438-238EA 601.4 CAD
77438-238
Anti-CYP 3A4, 3A7, 3A5, and 3A9 Rabbit Polyclonal Antibody
Antibodies
Cytochromes P450 are a group of heme thiolate monooxygenases. In liver microsomes, this enzyme is involved in an NADPH dependent electron transport pathway. It performs a variety of oxidation reactions (e.g. caffeine 8 oxidation, omeprazole sulphoxidation, midazolam 1 hydroxylation and midazolam 4 hydroxylation) of structurally unrelated compounds, including steroids, fatty acids, and xenobiotics. The enzyme also hydroxylates etoposide.

Type: Primary
Antigen: CYP 3A4, 3A7, 3A5, and 3A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR