PureCube Zr-NTA MagBeads

Supplier: CUBE BIOTECH

31501-ZR 31505-ZR 31525-ZR 31590-ZR
77393-568EA 202.65 CAD
77393-568 77393-580 77393-592 77393-604
PureCube Zr-NTA MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube Zr-NTA MagBeads/magnetic beads were developed for the enrichment of phosphorylated proteins. They can also be used for the purification of His-tagged proteins as a secondary function.


  • Usage: Enrichment of phosphopeptides, specific binding and purification of 6× His-tagged proteins
  • Specificity: Affinity to His-tagged proteins
  • Affinity to phosphorylated biomolecules
  • Bead ligand: Fe-NTA
  • Bead size: 40 µm
  • Filling quantity: Delivered as a 50% suspension
  • Matrix: 7.5% crosslinked agarose
  • Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
  • pH stability: 2 to 14
  • Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile


The affinity matrix is based on 6% cross-linked agarose that was magnetized. The bead ligand is Zr-NTA and the particle diameter is 30 µm. They are very homogeneous in size, yielding in a high degree of reproducibility. The phosphopeptide enrichment with Cube Biotech MagBeads can be automated as it has already been proven in this paper.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR