Mouse Recombinant MSP2N2 (from E. coli)
Supplier: CUBE BIOTECH
26586 26582
77392-792PK 4337.8 CAD
77392-792 77392-790
Mouse Recombinant MSP2N2 (from E. coli)
Proteins and Peptides
Membrane scaffold protein, mouse origin.
  • Extinction coefficient (in water) ε280: 36900 M⁻¹cm⁻¹ (Human origin)
  • Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
  • Affinity tag: none
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP2N2 creates nanodiscs of ~17 nm in diameter.