DIBMA 12 in HEPES, PureCube for protein solubilisation

Supplier: CUBE BIOTECH

18012 18031 18032
77392-474EA 831.78 CAD
77392-474 77392-480 77392-482
DIBMA 12 in HEPES, PureCube for protein solubilisation
DIBMA 12

With diisobutylene-maleic acid (DIBMA), you can directly extract membrane proteins from cells without an intermediate step of detergent solubilization, like with SDS, which would usually interfere with the protein's function. Another advantage of DIBMA as a tool for protein solubilization is the lack of an absorbance maxima at 280 nm. In comparison to SMAs, this would usually interfere with protein quantification, as aromatic amino acids absorb at the same spectrum. PureCube DIBMA is lyophilized from two different buffer solutions (HEPES or TRIS) to ensure a stable pH of 7.5, which is ideal for most protein solubilizations. A good publication to read even more details about DIBMA is Oluwole et al. 2017.


Usage: Protein solubilization
Formula Weight 12,000 g/mol
pH: 7.5 in buffer
dn/dc: 1.35 M-1
Solubility > 10 % in H2O
Absorbance at 280 nm: < 0.3 (1 % solution)
Mg2+ Tolerance: Depending on DIBMA product, Increased with less charged DIBMAs
Ca2+ Tolerance: Depending on DIBMA product, Increased with less charged DIBMAs


Caution: Shipping Temperature: Ambient temperature. Storage of lyophilized copolymer: -20°C for several years. Storage of dissolved copolymer: 2-8°C for several day.

CAS Number: 37199-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR