Anti-Klrg1 Hamster Monoclonal Antibody [clone: [2F1]]

Supplier: Prosci
98-965
77134-614EA 655.7 CAD
77134-614
Anti-Klrg1 Hamster Monoclonal Antibody [clone: [2F1]]
Antibodies
Type: Primary
Antigen: Klrg1
Clonality: Monoclonal
Clone: [2F1]
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR