Anti-CD8A Mouse Monoclonal Antibody [clone: [UCHT4]] (PE (Phycoerythrin)-Cy5®)

Supplier: Prosci
99-444
77132-892EA 703.1 CAD
77132-892
Anti-CD8A Mouse Monoclonal Antibody [clone: [UCHT4]] (PE (Phycoerythrin)-Cy5®)
Antibodies
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: [UCHT4]
Conjugation: PE (Phycoerythrin)-Cy5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rhesus Monkey, Cynomolgus Monkey, Chimpanzee
Order Now

SPECIFICATIONS

Antigen symbol CD8A
Antigen name T-cell surface glycoprotein CD8 alpha chain
Environmentally Preferable
Antigen synonyms T8|Tp32|Lyt2|Leu-2
Antibody type Primary
Clonality Monoclonal
Conjugation PE (Phycoerythrin)-Cy5®
Clone [UCHT4]
Reactivity
Host Mouse
Gene ID 925
Isotype IgG2a
Flow cytometry Yes
Modification None
Immunogen Thymocytes and Sézary T cells.
Form Liquid
Concentration Batch dependent
Storage buffer Supplied in PBS/NaN3 and a stabilizing agent.
Storage temperature Store vial at 2...8 °C.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR