Anti-IL5 Rat Monoclonal Antibody [clone: [JES1-39D10]]

Supplier: Prosci
99-660
77132-882EA 703.1 CAD
77132-882
Anti-IL5 Rat Monoclonal Antibody [clone: [JES1-39D10]]
Antibodies
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: [JES1-39D10]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR