Rhesus monkey Recombinant Rhesus macaque Complement Factor D/CFD Protein (from HEK293 cells)

Supplier: Prosci

11-333
77128-536EA 876.9 CAD
77128-536
Rhesus monkey Recombinant Rhesus macaque Complement Factor D/CFD Protein (from HEK293 cells)
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name CFD
Environmentally Preferable
Protein synonyms PFD|Complement factor D|CFD|DF|Adipsin
Protein/peptide type Recombinant
Species Rhesus monkey
Source HEK293 cells
Conjugation Unconjugated
Gene ID 102136571
Sequence Ile 26 - Ala 253
Purity >95% as determined by SDS-PAGE
Formulation Lyophilized. Tris with Glycine,  Arginine and NaCl,  pH 7.5.
Molecular weight 51.0 kDa
Storage conditions Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
Shipping temperature RT
Tested applications WB

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR