Rhcg Peptide

Supplier: Prosci

42-920P
77126-964EA 450.3 CAD
77126-964
Rhcg Peptide
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Rhcg
Environmentally Preferable
Protein synonyms Rh family type C glycoprotein|BB065800|Rhesus blood group family type C glycoprotein|Ammonium transporter Rh type C
Protein/peptide type Synthetic
Species
Source
Conjugation Unconjugated
Gene ID 56315
Formulation Lyophilized. Rhcg peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
Storage conditions Store at −20 °C. When reconstituted minimize freezing and thawing.
Shipping temperature RT
Tested applications Blocking

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR