Anti-Tnf Rat Monoclonal Antibody [clone: [MP6-XT3]] (Biotinylated)

Supplier: Prosci
99-714
77126-828EA 703.1 CAD
77126-828
Anti-Tnf Rat Monoclonal Antibody [clone: [MP6-XT3]] (Biotinylated)
Antibodies
Type: Primary
Antigen: Tnf
Clonality: Monoclonal
Clone: [MP6-XT3]
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR