Anti-Synaptotagmin-3 Goat Polyclonal Antibody

Supplier: Prosci
43-333
77126-198EA 583.02 CAD
77126-198
Anti-Synaptotagmin-3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Synaptotagmin-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR