Anti-H2-D1 Mouse Monoclonal Antibody [clone: [34-5-8S]]

Supplier: Prosci
99-080
77122-260EA 655.7 CAD
77122-260
Anti-H2-D1 Mouse Monoclonal Antibody [clone: [34-5-8S]]
Antibodies
Type: Primary
Antigen: H2-D1
Clonality: Monoclonal
Clone: [34-5-8S]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR