Anti-NPC1 Goat Polyclonal Antibody

Supplier: Prosci
43-348
77120-412EA 741.02 CAD
77120-412
Anti-NPC1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR