AIRE Peptide

Supplier: Prosci

45-225P
77120-120EA 450.3 CAD
77120-120
AIRE Peptide
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name AIRE
Protein synonyms APECED protein|AIRE1|APECED|Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein|PGA1|Autoimmune regulator|APSI|APS1
Environmentally Preferable
Protein/peptide type Synthetic
Species Human
Source
Conjugation Unconjugated
Gene ID 326
Formulation Lyophilized. AIRE peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
Storage conditions Store at −20 °C. When reconstituted minimize freezing and thawing.
Shipping temperature RT
Tested applications Blocking

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR