Anti-Cd38 Rat Monoclonal Antibody [clone: [NIMR-5]]

Supplier: Prosci
98-748
77119-470EA 703.1 CAD
77119-470
Anti-Cd38 Rat Monoclonal Antibody [clone: [NIMR-5]]
Antibodies
Type: Primary
Antigen: Cd38
Clonality: Monoclonal
Clone: [NIMR-5]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol Cd38
Environmentally Preferable
Antigen name CD38 antigen
Antigen synonyms T10|cyclic ADP-ribose hydrolase|ADP-ribosyl cyclase
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone [NIMR-5]
Reactivity Mouse
Host Rat
Gene ID 12494
Isotype IgG2a
ImmunoChemistry Yes
Flow cytometry Yes
Modification None
Immunogen BCL1 plasma membrane glycoproteins.
Form Liquid
Concentration 0.5 mg/ml
Storage buffer Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added.
Storage temperature Store vial at 2...8 °C.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR