Anti-ORF8 Rabbit Polyclonal Antibody

Supplier: Prosci
9289
77119-424EA 687.3 CAD
77119-424
Anti-ORF8 Rabbit Polyclonal Antibody
Antibodies
SARS-CoV-2 (COVID-19) ORF8 antibody can be stored at 4 to 730 °C for three months and –20 to +730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.

Type: Primary
Antigen: ORF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

SPECIFICATIONS

Antigen symbol ORF8
Antigen name ORF8 protein
Environmentally Preferable
Antigen synonyms ORF8|ns8|Non-structural protein 8|ORF8 protein
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Virus
Host Rabbit
Gene ID 43740577
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Anti-SARS-CoV-2 (COVID-19) ORF8 antibody 9289 was raised against a peptide corresponding to 13 amino acids near the central domain of SARS-CoV-2 (COVID-19) ORF8 protein. The immunogen is located within 30-80 amino acids of the SARS-CoV-2 (COVID-19) ORF8 protein.
Form Liquid
Purification SARS-CoV-2 (COVID-19) ORF8 Antibody is affinity chromatography purified via peptide column.
Concentration 1 mg/ml
Storage buffer SARS-CoV-2 (COVID-19) ORF8 antibody is supplied in PBS containing 0.02% sodium azide.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR