APM Adult Body Bag, 16 Mil PVC Laminated Polyester, Center/Envelope Zipper, 6 Handles, Black
Supplier: Mortech Manufacturing
BE801-BK-2XL BE801-BK-3XL BE801-BK-4XL BE801-BK-XL BE801E-BK BE801E-BK-2XL BE801E-BK-3XL BE801E-BK-4XL BE801E-BK-XL BE801-BK
76945-218CS 1528.76 CAD
76945-218 76945-220 76945-222 76945-224 76945-236 76945-238 76945-240 76945-242 76945-244 76945-216
APM Adult Body Bag, 16 Mil PVC Laminated Polyester, Center/Envelope Zipper, 6 Handles, Black
Bags Storage Bags
Mopec in partnership with American Professional Manufacturing provide excellent body bags.
  • Custom-made, lockable slider for easy opening and closing
  • HD nylon zipper, 100% sewn with nylon thread
  • Dielectrically sealed seams
  • Available with center or envelope zipper
  • 6 handles secured by heavy-duty steel rivets
  • Leak resistance
Materials: 16 mil PVC laminated polyester
Dimensions: Various sizes (Available in Bariatric XL, 2XL, 3XL and 4XL)
Order Now
SPECIFICATIONS
Environmentally Preferable