Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
Supplier: AAT BIOQUEST INC
17630
76779-040EA 348.09 CAD
76779-040
Helixyte™ Green Fluorimetric Total Nucleic Acid Quantitation Kit
Assays Nucleic Acid Assays
Helixyte™ Green fluorimetric total nucleic acid quantitation kit is optimized for measuring the total amounts of nucleic acids with a fluorescence microplate reader.
  • Optimized for microplate readers
  • Sensitive and indiscriminate binding
The kit has all the essential reagents, including Helixyte™ Green all reagent, dilution buffer, and pre-diluted DNA standards.
Packaging: Plastic vial.
Caution: For research use only.