Anti-MDX-1103 Recombinant Antibody

Supplier: Prosci
10-334
76764-908EA 782.1 CAD
76764-908
Anti-MDX-1103 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen synonyms CNTO 095
Antibody type Primary
Clonality Recombinant
Conjugation Unconjugated
Reactivity
Host
Isotype IgG1 kappa
Modification None
Immunogen Homo sapiens/ITGAV_ITGB3 [Homo sapiens].
Purification >95%
Concentration Batch dependent
Storage buffer PBS buffer pH 7.5 .
Storage temperature Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 ...730 °C for 1-2 weeks. Store at –20…+730 °C for 12 months. Store at –80…+730 °C for long term storage.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR