Anti-SUCLG1 Rabbit Polyclonal Antibody

Supplier: Prosci
16-002
76764-236EA 492.88 CAD
76764-236
Anti-SUCLG1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SUCLG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol SUCLG1
Environmentally Preferable
Antigen name Succinate-CoA ligase alpha subunit
Antigen synonyms SUCLA1|succinyl-CoA ligase [ADP/GDP-forming] subunit alpha|GALPHA|SCS-alpha|mitochondrial|succinate--CoA ligase [ADP/GDP-forming] subunit alpha|MTDPS9|succinyl-CoA ligase [GDP-forming] subunit alpha|succinyl-CoA synthetase subunit alpha
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 8802
Isotype IgG
Western blot Yes
Modification None
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 41-346 of human SUCLG1 (NP_003840.2).
Form Liquid
Purification Affinity purification
Concentration Batch dependent
Storage buffer PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
Storage temperature Store at −20 °C. Avoid freeze/thaw cycles.
Shipping temperature 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR