Anti-BAY-1093884 Recombinant Antibody

Supplier: Prosci
10-972
76763-430EA 782.1 CAD
76763-430
Anti-BAY-1093884 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-lambda
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR